Learn More
Cayman Chemical (E/Z)-4-hydroxy Tamoxifen, Purity: ≥98% (mixture of isomers), CAS: 68392-35-8, 50 mg
SDP
Catalog No. 502965465
Encompass_Preferred
A mixture of (E)-4-hydroxy tamoxifen and (Z)-4-hydroxy tamoxifen; (Z)-4-hydroxy Tamoxifen (100 nM) inhibits 17β-estradiol-induced increases in the mRNA encoding progesterone receptor in MCF-7 cancer cells at 100 nM; (Z)-4-hydroxy Tamoxifen inhibits the proliferation of MCF-7 and MDA-MB-231 cancer cells (IC50s = 27 and 18 µM, respectively)
Catalog No. 50-296-5465 Supplier Cayman Chemical Supplier No. 1730850 MG
A mixture of (E)-4-hydroxy tamoxifen and (Z)-4-hydroxy tamoxifen; (Z)-4-hydroxy Tamoxifen (100 nM) inhibits 17β-estradiol-induced increases in the mRNA encoding progesterone receptor in MCF-7 cancer cells at 100 nM; (Z)-4-hydroxy Tamoxifen inhibits the proliferation of MCF-7 and MDA-MB-231 cancer cells (IC50s = 27 and 18 µM, respectively)