Learn More
Research Products International Corp EZ-96 DNA Methylation-Lightning MagPrep, 4 x 96 Preps
SDP
Catalog No. 501977017
Encompass
The EZ-96 DNA Methylation-Lightning MagPrep Kit is the fastest magnetic bead-based DNA bisulfite conversion kit for high-throughput and automated methylation analysis. No preparation is necessary with the ready-to-use conversion reagent - simply add this unique reagent to a DNA sample and incubate the reaction for about one hour. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. This magnetic bead-based bisulfite conversion kit produces the highest yields of bisulfite-converted DNA that is ideal for PCR, arrays, library preps, and Next-Generation sequencing.
High-throughput procedure for automated rapid and complete bisulfite conversion of DNA for methylation analysis.
Ready-to-use conversion reagent is added directly to DNA.
High-yield, converted DNA is ideal for PCR, Methylation Specific PCR (MSP), arrays, library preps, Next-Generation sequencing, etc.
Catalog No. 50-197-7017 Supplier Research Products International Corp Supplier No. ZD5046
The EZ-96 DNA Methylation-Lightning MagPrep Kit is the fastest magnetic bead-based DNA bisulfite conversion kit for high-throughput and automated methylation analysis. No preparation is necessary with the ready-to-use conversion reagent - simply add this unique reagent to a DNA sample and incubate the reaction for about one hour. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. This magnetic bead-based bisulfite conversion kit produces the highest yields of bisulfite-converted DNA that is ideal for PCR, arrays, library preps, and Next-Generation sequencing.
High-throughput procedure for automated rapid and complete bisulfite conversion of DNA for methylation analysis.
Ready-to-use conversion reagent is added directly to DNA.
High-yield, converted DNA is ideal for PCR, Methylation Specific PCR (MSP), arrays, library preps, Next-Generation sequencing, etc.