Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Direct MagPrep (8x96 rxns.)
SDP
Catalog No. 502037130
Encompass_Preferred
The increased sensitivity of this kit makes it possible to amplify bisulfite converted DNA from as few as 10 cells or 50 pg DNA. These innovations have been coupled to a magnetic bead based clean-up for high-throughput methylation analysis. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. The kits have been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for PCR, array, bisulfite and next generation sequencing, etc.
Catalog No. 50-203-7130 Supplier Zymo Research Corporation Supplier No. D5045
The increased sensitivity of this kit makes it possible to amplify bisulfite converted DNA from as few as 10 cells or 50 pg DNA. These innovations have been coupled to a magnetic bead based clean-up for high-throughput methylation analysis. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. The kits have been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for PCR, array, bisulfite and next generation sequencing, etc.