Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Gold MagPrep (4x96 rxns.)
SDP
Catalog No. 502037128
Encompass_Preferred
The EZ-96 DNA Methylation-Gold MagPrep integrates DNA denaturation and bisulfite conversion processes into one step followed by a magnetic bead-based clean-up for high-throughput methylation analysis. To streamline the procedure, high temperature is used instead of sodium hydroxide to denature DNA. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. The EZ DNA Methylation-Gold kits have been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for downstream analyses, including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-203-7128 Supplier Zymo Research Corporation Supplier No. D5042
The EZ-96 DNA Methylation-Gold MagPrep integrates DNA denaturation and bisulfite conversion processes into one step followed by a magnetic bead-based clean-up for high-throughput methylation analysis. To streamline the procedure, high temperature is used instead of sodium hydroxide to denature DNA. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. The EZ DNA Methylation-Gold kits have been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for downstream analyses, including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.