Learn More
Research Products International Corp EZ-96 DNA Methylation Kit, Deep Well, 2 x 96 Rxns
SDP
Catalog No. 501977197
Encompass
The EZ-96 DNA Methylation Kits are bisulfite conversion kits that feature a high-throughput (96-well spin-plate), simplified procedure that streamlines bisulfite conversion of DNA. Cytosines undergo a three-step reaction with sodium bisulfite during which the cytosine is converted into uracil. The innovative in-column desulphonation technology eliminates otherwise cumbersome precipitations. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc. Note: Catalog # ZD5004 is recommended for use with the Illumina Infinium MethylationEPIC BeadChip array.
Streamlined, proven procedure for bisulfite conversion of DNA.
Desulphonation and recovery of bisulfite-treated DNA with a 96-well spin-column plate.
Catalog No. 50-197-7197 Supplier Research Products International Corp Supplier No. ZD5004
The EZ-96 DNA Methylation Kits are bisulfite conversion kits that feature a high-throughput (96-well spin-plate), simplified procedure that streamlines bisulfite conversion of DNA. Cytosines undergo a three-step reaction with sodium bisulfite during which the cytosine is converted into uracil. The innovative in-column desulphonation technology eliminates otherwise cumbersome precipitations. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc. Note: Catalog # ZD5004 is recommended for use with the Illumina Infinium MethylationEPIC BeadChip array.
Streamlined, proven procedure for bisulfite conversion of DNA.
Desulphonation and recovery of bisulfite-treated DNA with a 96-well spin-column plate.