Learn More
Zymo Research Corporation EZ-96 DNA Methylation MagPrep (8x96 rxns.)
SDP
Catalog No. 502037126
Encompass_Preferred
The EZ-96 DNA Methylation MagPrep features a high-throughput (96-well), simplified procedure that streamlines bisulfite conversion of DNA. The kit is based on the three-step reaction that takes place between cytosine and sodium bisulfite during which cytosine is converted into uracil. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-203-7126 Supplier Zymo Research Corporation Supplier No. D5041
The EZ-96 DNA Methylation MagPrep features a high-throughput (96-well), simplified procedure that streamlines bisulfite conversion of DNA. The kit is based on the three-step reaction that takes place between cytosine and sodium bisulfite during which cytosine is converted into uracil. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.