Learn More
Cayman Chemical (E/Z)-Ajoene, Purity: ≥95%, CAS: 92285-01-3, 10 mg
SDP
Catalog No. 502910709
Encompass_Preferred
A disulfide with antitumor and antiplatelet activities; cytotoxic to a variety of cancer cells (IC50s = 7-38 µM); reduces tumor growth and decreases the number of lung metastases in murine models of melanoma at 25 mg/kg; prevents thrombus formation in heparinized animals in a porcine model of severe vascular damage at 25 mg/kg
Catalog No. 50-291-0709 Supplier Cayman Chemical Supplier No. 3488110 MG
A disulfide with antitumor and antiplatelet activities; cytotoxic to a variety of cancer cells (IC50s = 7-38 µM); reduces tumor growth and decreases the number of lung metastases in murine models of melanoma at 25 mg/kg; prevents thrombus formation in heparinized animals in a porcine model of severe vascular damage at 25 mg/kg