Learn More
Euthanex The EZ-AF9000 Auto Flow Anesthesia System with Regulator, Induction Chamber, Beds, Breathers and Charcoal Filters
Catalog No. NC0933627
Encompass_Preferred
The EZ-AF9000 Auto Flow System is the most advanced anesthesia system available and offers high efficient, precise gas flow that is easy to use. The system has an automatic five output manifold that does not require flowmeters. Features include: Fixed flow rates are set specifically for mice and rats; Control each output individually with simple on/off switches; Five outputs supply anesthesia to the induction chamber and up to four breathers; Preset flow rates eliminate need for flowmeters and manual adjustments; Easily follow Protocols without calculations; Minimum opportunity for operator error; Flow rate is not impacted by the type or number of accessories connected; Precise, low flow gas saves money and protects personnel; Pushbutton oxygen flush purges induction chamber before removing animals; Carry handle facilitates safe, easy transport of the unit.
Catalog No. NC0933627 Supplier Euthanex Supplier No. EZAF9000
Add to Cart
Add to Cart
Request a Quote
The EZ-AF9000 Auto Flow System is the most advanced anesthesia system available and offers high efficient, precise gas flow that is easy to use. The system has an automatic five output manifold that does not require flowmeters. Features include: Fixed flow rates are set specifically for mice and rats; Control each output individually with simple on/off switches; Five outputs supply anesthesia to the induction chamber and up to four breathers; Preset flow rates eliminate need for flowmeters and manual adjustments; Easily follow Protocols without calculations; Minimum opportunity for operator error; Flow rate is not impacted by the type or number of accessories connected; Precise, low flow gas saves money and protects personnel; Pushbutton oxygen flush purges induction chamber before removing animals; Carry handle facilitates safe, easy transport of the unit.