Learn More
EZ BIOSYSTEMS INC EZ Biosystems Transfection Reagent, J774A.1 Cell, Mouse Reticulum Cell Sarcoma, Avalanche™
Catalog No. NC3988608
Encompass
This proprietary transfection reagent is specifically designed for J774A.1 Cell Avalanche™ Transfection Reagent, offering high efficiency and cell viability. Its unique formulation supports delivery of various nucleic acids for molecular biology research.
  • Broad spectrum nucleic acid transfection, including plasmids, mRNA, and siRNA.
  • Optimized formulation for J774A.1 cells.
  • Gentle to cells, ensuring high viability.
  • Economical, requiring less nucleic acid and reagent.
  • Delivers single or multiple plasmids.
  • Animal-free.
  • Compatible with serum and various plate formats.
  • Forms uniformly sized complexes for reproducible results.
  • Also highly effective on RAW 264.7, 786-O, Caki-1, MDCK, Vero, 293, and 293T/17 cells.
Catalog No. NC3988608 Supplier EZ BIOSYSTEMS INC Supplier No. EZTJ7741
This proprietary transfection reagent is specifically designed for J774A.1 Cell Avalanche™ Transfection Reagent, offering high efficiency and cell viability. Its unique formulation supports delivery of various nucleic acids for molecular biology research.
  • Broad spectrum nucleic acid transfection, including plasmids, mRNA, and siRNA.
  • Optimized formulation for J774A.1 cells.
  • Gentle to cells, ensuring high viability.
  • Economical, requiring less nucleic acid and reagent.
  • Delivers single or multiple plasmids.
  • Animal-free.
  • Compatible with serum and various plate formats.
  • Forms uniformly sized complexes for reproducible results.
  • Also highly effective on RAW 264.7, 786-O, Caki-1, MDCK, Vero, 293, and 293T/17 cells.