Learn More
Scytek Laboratories EZ BLOCK (500 ML) EBZ500
SDP
Catalog No. NC1855738
Encompass_Preferred
EZ Block has been developed to use with immunolabeling techniques for the reduction of nonspecific background staining, and simultaneously reducing the handling of animal serums in the laboratory. The need to match species with the secondary antibody is eliminated due to the lack of normal serum in this product. EZ Block has been shown to be effective for immunohistochemical, ELISA, blot and In-situ techniques and requires no mixing or diluting.
Catalog No. NC1855738 Supplier Scytek Laboratories Supplier No. EBZ500
EZ Block has been developed to use with immunolabeling techniques for the reduction of nonspecific background staining, and simultaneously reducing the handling of animal serums in the laboratory. The need to match species with the secondary antibody is eliminated due to the lack of normal serum in this product. EZ Block has been shown to be effective for immunohistochemical, ELISA, blot and In-situ techniques and requires no mixing or diluting.