Learn More
LAMDA BIOTECH INC EZ-Blue Protein Gel Staining
Catalog No. NC3724891
Encompass_Preferred
EZ-Blue™ is an advanced Coomassie-based protein staining solution engineered for simplified, single-step gel visualization. This ready-to-use formulation eliminates the need for fixing, washing, or destaining steps while delivering high-sensitivity protein detection.
Catalog No. NC3724891 Supplier LAMDA BIOTECH INC Supplier No. GPS002
EZ-Blue™ is an advanced Coomassie-based protein staining solution engineered for simplified, single-step gel visualization. This ready-to-use formulation eliminates the need for fixing, washing, or destaining steps while delivering high-sensitivity protein detection.