Learn More
Apexbio Technology LLC EZ Cap™ Cas9 5-moUTP, 5x1mg (1mg/mL)
SDP
Catalog No. 501998330
Encompass_Preferred
The Cas9/guide RNA (Cas9/gRNA) system is commonly used for genome editing. Upon delivery of these components to the nucleus of a cell, the RNA guide sequence targets the site of interest and the Cas9 protein is employed to perform the DNA cleavage. 5moUTP-modified Cas9 mRNA is capped using Cap 1, a novel co-transcriptional capping method, which yields natural Cap 1 with high capping efficiency. 5moUTP modification reduces innate immune responses and increases Cas9 activity. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8330 Supplier Apexbio Technology LLC Supplier No. R10155
The Cas9/guide RNA (Cas9/gRNA) system is commonly used for genome editing. Upon delivery of these components to the nucleus of a cell, the RNA guide sequence targets the site of interest and the Cas9 protein is employed to perform the DNA cleavage. 5moUTP-modified Cas9 mRNA is capped using Cap 1, a novel co-transcriptional capping method, which yields natural Cap 1 with high capping efficiency. 5moUTP modification reduces innate immune responses and increases Cas9 activity. Other sizes are also available. Please inqury us for quote.