Learn More
Apexbio Technology LLC EZ Cap™ Cre mRNA (5-moUTP) 1mg (1mg/mL)
SDP
Catalog No. 5000096481
Encompass_Preferred
EZ Cap Cre mRNA 5-moUTP encodes Cre recombinase a tyrosine recombinase enzyme that facilitates homologous DNA recombination between two specific loxP recognition sites It is designed to promote precise DNA recombination thereby enabling conditional gene modification in mammalian systems EZ Cap Cre mRNA 5-moUTP exerts its biological activity primarily through expression of Cre recombinase which catalyzes site-specific recombination at loxP sites This mRNA incorporates a Cap 1 structure and 5-methoxyuridine triphosphate (5-moUTP) modification to reduce innate immune responses and improve translational stability Based on these properties EZ Cap Cre mRNA 5-moUTP holds research potential in genome editing experiments conditional gene knockout studies and reporter gene activation in mammalian research models
Catalog No. 50-000-96481 Supplier Apexbio Technology LLC Supplier No. R10461
EZ Cap Cre mRNA 5-moUTP encodes Cre recombinase a tyrosine recombinase enzyme that facilitates homologous DNA recombination between two specific loxP recognition sites It is designed to promote precise DNA recombination thereby enabling conditional gene modification in mammalian systems EZ Cap Cre mRNA 5-moUTP exerts its biological activity primarily through expression of Cre recombinase which catalyzes site-specific recombination at loxP sites This mRNA incorporates a Cap 1 structure and 5-methoxyuridine triphosphate (5-moUTP) modification to reduce innate immune responses and improve translational stability Based on these properties EZ Cap Cre mRNA 5-moUTP holds research potential in genome editing experiments conditional gene knockout studies and reporter gene activation in mammalian research models