Learn More
Apexbio Technology LLC EZ Cap™ EGFP mRNA (5-moUTP), 5x1mg (1mg/mL)
SDP
Catalog No. 501998493
Encompass_Preferred
This product has EGFP mRNA with Cap 1 structure and contains 5-moUTP modification, which can provide higher transcription efficiency and inhibit RNA-mediated innate immune activation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8493 Supplier Apexbio Technology LLC Supplier No. R10165
This product has EGFP mRNA with Cap 1 structure and contains 5-moUTP modification, which can provide higher transcription efficiency and inhibit RNA-mediated innate immune activation. Other sizes are also available. Please inqury us for quote.