Learn More
Apexbio Technology LLC EZ-CAP-EGFP-MRNA-5X1MG
SDP
Catalog No. 5000570707
Encompass_Preferred
EZ Cap EGFP mRNA is a modified messenger RNA commonly used in biomedical research specifically within the fields of gene expression and cellular imaging Originating from enhanced green fluorescent protein (EGFP) it acts by delivering the genetic blueprint for EGFP synthesis directly into cells allowing for real-time visualization of cellular processes through fluorescence The mRNA is efficiently translated in vitro displaying activity in nanomolar to low micromolar ranges Applications often involve various cell models including primary cells and cancer cell lines as well as animal studies where it aids in observing gene expression patterns and assessing transfection efficiency It is also valuable in drug screening and therapeutic development particularly in verifying delivery systems and cellular uptake Experimental concentrations typically depend on the specific research design ensuring adaptability to diverse scientific inquiries
Catalog No. 50-005-70707 Supplier Apexbio Technology LLC Supplier No. R10025
EZ Cap EGFP mRNA is a modified messenger RNA commonly used in biomedical research specifically within the fields of gene expression and cellular imaging Originating from enhanced green fluorescent protein (EGFP) it acts by delivering the genetic blueprint for EGFP synthesis directly into cells allowing for real-time visualization of cellular processes through fluorescence The mRNA is efficiently translated in vitro displaying activity in nanomolar to low micromolar ranges Applications often involve various cell models including primary cells and cancer cell lines as well as animal studies where it aids in observing gene expression patterns and assessing transfection efficiency It is also valuable in drug screening and therapeutic development particularly in verifying delivery systems and cellular uptake Experimental concentrations typically depend on the specific research design ensuring adaptability to diverse scientific inquiries