Learn More
Apexbio Technology LLC EZ CAP FIREFLY LUC MRNA-1mg
SDP
Catalog No. 5000897664
Encompass_Preferred
Enhanced EZ Cap Firefly Luciferase mRNA is widely used in biomedical research as a reporter gene tool to study gene expression and signal transduction pathways Originating as a synthetic mRNA it encodes luciferase an enzyme that catalyzes light production enabling quantitative measurement of transcriptional activity Its mechanism involves transcriptional activation that results in bioluminescence providing a sensitive and quantifiable readout In vitro it demonstrates activity in the nanomolar to low micromolar range making it highly sensitive for detecting gene expression levels Common applications include use in cell models for investigating gene function in animal studies for monitoring in vivo gene expression and in high-throughput drug screening assays to evaluate potential therapeutic compounds Experimental concentrations typically depend on the specific research design allowing flexibility across various experimental setups
Catalog No. 50-008-97664 Supplier Apexbio Technology LLC Supplier No. R11051
Enhanced EZ Cap Firefly Luciferase mRNA is widely used in biomedical research as a reporter gene tool to study gene expression and signal transduction pathways Originating as a synthetic mRNA it encodes luciferase an enzyme that catalyzes light production enabling quantitative measurement of transcriptional activity Its mechanism involves transcriptional activation that results in bioluminescence providing a sensitive and quantifiable readout In vitro it demonstrates activity in the nanomolar to low micromolar range making it highly sensitive for detecting gene expression levels Common applications include use in cell models for investigating gene function in animal studies for monitoring in vivo gene expression and in high-throughput drug screening assays to evaluate potential therapeutic compounds Experimental concentrations typically depend on the specific research design allowing flexibility across various experimental setups