Learn More
Apexbio Technology LLC EZ CAP LUC MRNA 5MOU-5mg
SDP
Catalog No. 5000897667
Encompass_Preferred
Enhanced EZ Cap Firefly Luciferase mRNA (5-moUTP) is a cutting-edge tool in the field of molecular biology specifically used for gene expression analysis Originating from advancements in mRNA technology it operates by encoding the firefly luciferase enzyme which emits bioluminescence upon substrate interaction allowing researchers to study gene expression pathways with high sensitivity It demonstrates common in vitro activity in the nanomolar to low micromolar range This mRNA is widely used in various applications such as cell models animal studies and high-throughput drug screening to monitor gene expression and cellular responses Experimental concentrations typically depend on the specific research design enabling tailored approaches depending on the study objectives This versatile tool enhances the monitoring of biological processes in research contributing significantly to advancements in gene expression studies and therapeutic developments
Catalog No. 50-008-97667 Supplier Apexbio Technology LLC Supplier No. R11065
Enhanced EZ Cap Firefly Luciferase mRNA (5-moUTP) is a cutting-edge tool in the field of molecular biology specifically used for gene expression analysis Originating from advancements in mRNA technology it operates by encoding the firefly luciferase enzyme which emits bioluminescence upon substrate interaction allowing researchers to study gene expression pathways with high sensitivity It demonstrates common in vitro activity in the nanomolar to low micromolar range This mRNA is widely used in various applications such as cell models animal studies and high-throughput drug screening to monitor gene expression and cellular responses Experimental concentrations typically depend on the specific research design enabling tailored approaches depending on the study objectives This versatile tool enhances the monitoring of biological processes in research contributing significantly to advancements in gene expression studies and therapeutic developments