Learn More
Marketlab, Inc. EZ-Cart Double Basket Cart
Description
- Outfit with helpful, interchangeable RediCart accessories
- 200 lb. total weight capacity (100 lb. per shelf)
- 4 in. non-marking wheels (2 swivel, 2 tracking)
- Powder-coated steel finish
- Baskets corral smaller items
- Integrated hooks for hanging items
- Excellent maneuverability in tight spaces
Specifications
Specifications
| Product Type | Basket Cart |
| Capacity (Metric) | 90.7185 kg |
| Material | Powder-coated Steel |