Learn More
Cole-Parmer Diba Omnifit 006EZ-05-10-AFEZ, Chromatography Column with 1 Fixed & 1 Adjustable Endpiece, 5 mm ID x 100 mm L
Supplier:  Cole-Parmer 1194276
Encompass_Preferred
Ergonomically designed endpieces provide simple and quick height adjustmentPEEK column threads offer outstanding durabilityPrecision borosilicate glass columns provide inertness, visibility, and performanceSuperior frit placement adds bed support and even distribution of sample flowReusable columns let you pack your own media specific to your application
Catalog No. NC2823675