Learn More
Cole-Parmer Diba Omnifit EZ Chromatography Column w/ 2 Fixed Endpieces, 10 x 100 mm; 1/ea
Catalog No. NC1475203 Shop All Cole-Parmer Products
Encompass
Ergonomically designed EZ columns are easy to use yet provide excellent results. Columns are ideal for low- to medium-pressure applications and are resistant to many solvents used in liquid chromatography. Modular design gives you the flexibility to mix and match column sizes with different endpiece types to meet your specific requirements. Columns come with one fixed and one adjustable endpieces pre-assembled with 25 µm PE frits. Each column is supplied assembled and ready to use, with all the tubing, fittings, and connections needed to get started.
Catalog No. NC1475203 Supplier Cole-Parmer Supplier No. 1194310
Ergonomically designed EZ columns are easy to use yet provide excellent results. Columns are ideal for low- to medium-pressure applications and are resistant to many solvents used in liquid chromatography. Modular design gives you the flexibility to mix and match column sizes with different endpiece types to meet your specific requirements. Columns come with one fixed and one adjustable endpieces pre-assembled with 25 µm PE frits. Each column is supplied assembled and ready to use, with all the tubing, fittings, and connections needed to get started.