Learn More
Med Vet International EZ Circuit Cleaner Connector, J0248FX, Each
Supplier:  Med Vet International JOR248FX
Encompass_Preferred
PROPERLY CLEANS AND DISINFECTS BETWEEN PATIENTS – SAFELY AND QUICKLY Unilimb or F Circuits are a popular anesthesia circuit in veterinary medicine. A challenge in their use is proper cleaning and disinfection between patients. With the EZ Circuit Cleaner Connector, one to five anesthesia circuits can now be cleaned safely and quickly by utilizing warm air when connecting directly to a compatible warm air unit. - Hooks the anesthesia circuit on one end and the hose of a warm air unit on the other end - Utilizes a warm air unit, to direct warm drying air down the dual tubes of a Unilimb or F Circuit - Five breathing circuits can be connected simultaneously by the removal of black capsNot compatible with every warm air unit, i.e. Bair HuggerOnly 3 black plugs are included with connector.
Catalog No. 50-005-82545