Learn More
Apexbio Technology LLC EZ Cap CM-EGFP Probe mRNA (m1Ψ) 100ug (1mg/mL)
SDP
Catalog No. 503107513
Encompass_Preferred
CM-EGFP mRNA encodes a fusion protein comprised of a cell membrane (CM) targeting signal sequence and enhanced green fluorescent protein (EGFP) By fusing EGFP with a membrane localization signal this probe enables real-time visualization of dynamic changes occurring at the cell membrane of living cells particularly beneficial for research involving cell membrane-related processes such as signal transduction membrane protein interactions and alterations in membrane structure EZ Cap CM-EGFP Probe mRNA (m1 ) is provided at 1 mg/mL It is specifically designed for cell membrane-specific expression of a green fluorescent protein label This mRNA is synthesized through co-transcriptional capping resulting in a Cap 1 structure The incorporation of m1 modifications poly(A) tail significantly enhances mRNA stability and lifetime both in vitro and in vivo
Catalog No. 50-310-7513 Supplier Apexbio Technology LLC Supplier No. R11190.1
CM-EGFP mRNA encodes a fusion protein comprised of a cell membrane (CM) targeting signal sequence and enhanced green fluorescent protein (EGFP) By fusing EGFP with a membrane localization signal this probe enables real-time visualization of dynamic changes occurring at the cell membrane of living cells particularly beneficial for research involving cell membrane-related processes such as signal transduction membrane protein interactions and alterations in membrane structure EZ Cap CM-EGFP Probe mRNA (m1 ) is provided at 1 mg/mL It is specifically designed for cell membrane-specific expression of a green fluorescent protein label This mRNA is synthesized through co-transcriptional capping resulting in a Cap 1 structure The incorporation of m1 modifications poly(A) tail significantly enhances mRNA stability and lifetime both in vitro and in vivo