Learn More
Hach Company Hach ez COD Recycling Service (20-gallon)
Supplier:  Hach Company 2895420
Encompass
The ez COD Recycling Service provides a compliant collection, pickup, and recycling/disposal solution for used COD vials. The 20-gallon program includes container supply, scheduled pick-up, regulatory documentation, and final disposal handled by authorized waste processors.
  • 20-gallon container holds approximately 1,600 16 mm COD vials or 2,600 13 mm TNTplus vials.
  • Includes scheduled pick-up, documentation, and disposal/recycling services.
  • Intended for mercury-based COD products; not for mercury-free COD products.
  • Available to customers in the 48 contiguous United States.
  • Reduces on-site hazardous waste accumulation and simplifies regulatory recordkeeping.
Catalog No. NC3793035