Learn More
Euthanex The EZ-1130 Connection Kit with EZ-S2F Female Quick Disconnect
Catalog No. NC9992309
Encompass_Preferred
The EZ-1130 Connection Kit is required to integrate third party equipment into an E-Z Anesthesia system. It includes one EZ-S2F Female Quick Disconnect with 6 feet of "" ID tubing and clamps.
Catalog No. NC9992309 Supplier Euthanex Supplier No. EZ1130
 Restricted Item
Add to Cart
Add to Cart
The EZ-1130 Connection Kit is required to integrate third party equipment into an E-Z Anesthesia system. It includes one EZ-S2F Female Quick Disconnect with 6 feet of "" ID tubing and clamps.