Learn More
Apexbio Technology LLC EZ Cap Cre mRNA (ΨUTP) 5x1mg (1mg/mL)
SDP
Catalog No. 503107168
Encompass_Preferred
Cre recombinase is a tyrosine recombinase that recognizes and catalyzes homologous recombination between two loxP DNA sites This mRNA expresses Cre recombinase and is synthesized using co-transcriptional capping to generate a Cap 1 structure making it more suitable for mammalian systems and providing higher expression efficiency Additionally the mRNA is polyadenylated and incorporates pseudouridine ( UTP) modifications which help reduce host cell immune responses and enhance mRNA stability and translation efficiency
Catalog No. 50-310-7168 Supplier Apexbio Technology LLC Supplier No. R10295
Cre recombinase is a tyrosine recombinase that recognizes and catalyzes homologous recombination between two loxP DNA sites This mRNA expresses Cre recombinase and is synthesized using co-transcriptional capping to generate a Cap 1 structure making it more suitable for mammalian systems and providing higher expression efficiency Additionally the mRNA is polyadenylated and incorporates pseudouridine ( UTP) modifications which help reduce host cell immune responses and enhance mRNA stability and translation efficiency