Learn More
Electron Microscopy Sciences EZ Crimper™ Ez Crimper, Flipcap 13 mm Seals
SDP
Catalog No. 501898961
Encompass_Preferred
An adjustable stopper allows the user to determine the tightness of the seal while ensuring consistent crimping. The jaws are tapered to allow for the crimping of the vials in the racks. Ergonomic, PVC grip and spring assembly reduces hand fatigue. Attaches 13mm flip top seals.
Catalog No. 50-189-8961 Supplier Electron Microscopy Sciences Supplier No. 6033513
An adjustable stopper allows the user to determine the tightness of the seal while ensuring consistent crimping. The jaws are tapered to allow for the crimping of the vials in the racks. Ergonomic, PVC grip and spring assembly reduces hand fatigue. Attaches 13mm flip top seals.