Learn More
Electron Microscopy Sciences EZ Crimper™ Ez Crimper for Flipcap, 13 mm Seal
SDP
Catalog No. 501898962
Encompass_Preferred
An adjustable stopper allows the user to determine the tightness of the seal while ensuring consistent crimping. The jaws are tapered to allow for the crimping of the vials in the racks. Ergonomic, PVC grip and spring assembly reduces hand fatigue. Attaches 20mm flip top seals.
Catalog No. 50-189-8962 Supplier Electron Microscopy Sciences Supplier No. 6033520
An adjustable stopper allows the user to determine the tightness of the seal while ensuring consistent crimping. The jaws are tapered to allow for the crimping of the vials in the racks. Ergonomic, PVC grip and spring assembly reduces hand fatigue. Attaches 20mm flip top seals.