Learn More
Zymo Research Corporation EZ DNA Methlyation-Startup™ Kit (50 Preps) [Includes Room Temp Item D5020 x 1 and Dry Ice Items D5011 x 1 & E2003 x 1]
SDP
Catalog No. 50444334
Encompass_Preferred
The EZ DNA Methylation-Startup Kit provides the necessary tools for complete bisulfite conversion of DNA for methylation analysis. This kit includes reagents that allow for bisulfite conversion of purified DNA and DNA directly from blood, cells, and fresh or FFPE tissues without the prerequisite for upstream DNA purification. A fully methylated Universal Methylated Human DNA Standard is provided with a special primer set for PCR to assess conversion efficiency. Finally, a unique ZymoTaq DNA Polymerase is included for robust amplification of bisulfite-treated DNA.
Catalog No. 50-444-334 Supplier Zymo Research Corporation Supplier No. D5024
The EZ DNA Methylation-Startup Kit provides the necessary tools for complete bisulfite conversion of DNA for methylation analysis. This kit includes reagents that allow for bisulfite conversion of purified DNA and DNA directly from blood, cells, and fresh or FFPE tissues without the prerequisite for upstream DNA purification. A fully methylated Universal Methylated Human DNA Standard is provided with a special primer set for PCR to assess conversion efficiency. Finally, a unique ZymoTaq DNA Polymerase is included for robust amplification of bisulfite-treated DNA.