Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Direct MagPrep (4x96 rxns.)
SDP
Catalog No. 501251553
Encompass_Preferred
The EZ-96 DNA Methylation-Direct MagPrep allows simple and reliable DNA bisulfite conversion directly from blood, tissue, and cells without the prerequisite for DNA purification. The increased sensitivity of this kit makes it possible to amplify bisulfite-converted DNA from as few as 10 cells or 50 pg DNA. These innovations provide high-throughput methylation analysis. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. This kit has been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for downstream analyses, including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-125-1553 Supplier Zymo Research Corporation Supplier No. D5044
The EZ-96 DNA Methylation-Direct MagPrep allows simple and reliable DNA bisulfite conversion directly from blood, tissue, and cells without the prerequisite for DNA purification. The increased sensitivity of this kit makes it possible to amplify bisulfite-converted DNA from as few as 10 cells or 50 pg DNA. These innovations provide high-throughput methylation analysis. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. This kit has been designed to minimize template degradation, loss of DNA during treatment and clean-up, and to provide complete conversion of unmethylated cytosines. Recovered DNA is ideal for downstream analyses, including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.