Learn More
Zymo Research Corporation EZ DNA Methylation-Gold™ Kit (200 Rxns)
SDP
Catalog No. 50444298
Encompass_Preferred
The EZ DNA Methylation-Gold Kit is a refinement of our popular EZ DNA Methylation kit and uses heat denaturation instead of chemical denaturation of the input DNA. This allows for the denaturation and bisulfite conversion steps to be consolidated into one step, leading to a much-reduced incubation time. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-444-298 Supplier Zymo Research Corporation Supplier No. D5006
The EZ DNA Methylation-Gold Kit is a refinement of our popular EZ DNA Methylation kit and uses heat denaturation instead of chemical denaturation of the input DNA. This allows for the denaturation and bisulfite conversion steps to be consolidated into one step, leading to a much-reduced incubation time. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.