Learn More
Zymo Research Corporation EZ DNA Methylation-Gold™ Kit (50 Rxns)
SDP
Catalog No. 50444294
Encompass_Preferred
The EZ DNA Methylation-Gold Kit is a refinement of our popular EZ DNA Methylation kit and uses heat denaturation instead of chemical denaturation of the input DNA. This allows for the denaturation and bisulfite conversion steps to be consolidated into one step, leading to a much-reduced incubation time. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-444-294 Supplier Zymo Research Corporation Supplier No. D5005
The EZ DNA Methylation-Gold Kit is a refinement of our popular EZ DNA Methylation kit and uses heat denaturation instead of chemical denaturation of the input DNA. This allows for the denaturation and bisulfite conversion steps to be consolidated into one step, leading to a much-reduced incubation time. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.