Learn More
Research Products International Corp EZ DNA Methylation-Gold Kit, 50 Rxns
SDP
Catalog No. 501977606
Encompass
The EZ DNA Methylation-Gold Kit is a refinement of our popular EZ DNA Methylation kit and uses heat denaturation instead of chemical denaturation of the input DNA. This allows for the denaturation and bisulfite conversion steps to be consolidated into one step, leading to a much-reduced incubation time. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Complete bisulfite conversion of DNA in less than 3 hours.
Desulphonation and recovery of bisulfite-treated DNA with a spin column.
Recovered DNA is ideal for downstream analyses such as PCR, endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-197-7606 Supplier Research Products International Corp Supplier No. ZD5005
The EZ DNA Methylation-Gold Kit is a refinement of our popular EZ DNA Methylation kit and uses heat denaturation instead of chemical denaturation of the input DNA. This allows for the denaturation and bisulfite conversion steps to be consolidated into one step, leading to a much-reduced incubation time. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Complete bisulfite conversion of DNA in less than 3 hours.
Desulphonation and recovery of bisulfite-treated DNA with a spin column.
Recovered DNA is ideal for downstream analyses such as PCR, endonuclease digestion, sequencing, microarrays, etc.