Learn More
Zymo Research Corporation EZ DNA Methylation™ Kit (50 Rxns)
SDP
Catalog No. NC9038184
Encompass_Preferred
The EZ DNA Methylation Kit features a simplified procedure that streamlines bisulfite treatment of DNA. The EZ DNA Methylation Kit is based on the three-step reaction that takes place between cytosine and sodium bisulfite during which cytosine is converted into uracil. Innovative desulphonation technologies eliminate otherwise cumbersome precipitations. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including library preparation for Next-generation sequencing, PCR amplification, endonuclease digestion, sequencing, microarrays, etc. The EZ DNA Methylation Kit is recommended for use with Illumina Infinium MethylationEPIC BeadChip array.
Catalog No. NC9038184 Supplier Zymo Research Corporation Supplier No. D5001
The EZ DNA Methylation Kit features a simplified procedure that streamlines bisulfite treatment of DNA. The EZ DNA Methylation Kit is based on the three-step reaction that takes place between cytosine and sodium bisulfite during which cytosine is converted into uracil. Innovative desulphonation technologies eliminate otherwise cumbersome precipitations. The kit is designed to reduce template degradation and minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for downstream analyses including library preparation for Next-generation sequencing, PCR amplification, endonuclease digestion, sequencing, microarrays, etc. The EZ DNA Methylation Kit is recommended for use with Illumina Infinium MethylationEPIC BeadChip array.