Learn More
Research Products International Corp EZ DNA Methylation-Lightning Kit, 50 Reactions
SDP
Catalog No. 501977470
Encompass
The EZ DNA Methylation-Lightning Kit can rapidly bisulfite convert and purify DNA in less than 1.5 hours. A ready-to-use conversion reagent streamlines this process – simply add the reagent directly to the sample and incubate. Desulphonation and clean-up of the converted DNA is performed on a spin-column, allowing an elution volume as low as 10 µl. The kit achieves high conversion efficiency of unmodified cytosines into uracil, ensuring accurate downstream methylation analysis.
Fastest bisulfite conversion kit for complete bisulfite conversion of DNA for methylation analysis.
Ready-to-use conversion reagent is added directly to DNA.
High-yield, converted DNA is ideal for PCR, Methylation Specific PCR (MSP), arrays, library preps, Next-Generation sequencing, etc.
Catalog No. 50-197-7470 Supplier Research Products International Corp Supplier No. ZD5030
The EZ DNA Methylation-Lightning Kit can rapidly bisulfite convert and purify DNA in less than 1.5 hours. A ready-to-use conversion reagent streamlines this process – simply add the reagent directly to the sample and incubate. Desulphonation and clean-up of the converted DNA is performed on a spin-column, allowing an elution volume as low as 10 µl. The kit achieves high conversion efficiency of unmodified cytosines into uracil, ensuring accurate downstream methylation analysis.
Fastest bisulfite conversion kit for complete bisulfite conversion of DNA for methylation analysis.
Ready-to-use conversion reagent is added directly to DNA.
High-yield, converted DNA is ideal for PCR, Methylation Specific PCR (MSP), arrays, library preps, Next-Generation sequencing, etc.