Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Gold™ Kit (Deep-Well) (2 x 96 Rxns)
SDP
Catalog No. 501251524
Encompass_Preferred
The EZ-96 DNA Methylation-Gold™ Kit is a refinement of our popular EZ DNA Methylation™ kits. These products consolidate DNA denaturation and bisulfite conversion processes into one step, leading to a much faster bisulfite conversion. This is accomplished using temperature denaturation instead of chemical denaturation which is used in the original EZ DNA Methylation™ Kit. Also, the EZ DNA Methylation™-Gold™ Kits have been streamlined for high yield recovery of DNA following bisulfite treatment. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-125-1524 Supplier Zymo Research Corporation Supplier No. D5008
The EZ-96 DNA Methylation-Gold™ Kit is a refinement of our popular EZ DNA Methylation™ kits. These products consolidate DNA denaturation and bisulfite conversion processes into one step, leading to a much faster bisulfite conversion. This is accomplished using temperature denaturation instead of chemical denaturation which is used in the original EZ DNA Methylation™ Kit. Also, the EZ DNA Methylation™-Gold™ Kits have been streamlined for high yield recovery of DNA following bisulfite treatment. Recovered bisulfite-converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.