Learn More
Euthanex EZ-DOCK SMARTBOX Euthanasia System for use with Allentown NexGen Max High Density Mouse Cages. Connect the CO2 hose from regulator, push-in cage and system automatically begins and ends in compliance with AVMA Guidelines.
Catalog No. NC2259355
Encompass
Fits Allentown Nexgen Max cages only. The EZ-Dock combines the best features of all our systems. A built-in flow circuit and times are already preset for the specific cage type to ensure AMVA compliancy. Plug in, connect the provided gas line and the system is ready for use! Once the cage is placed in dock properly. The cycles begin and shut offs when time has elapsed. The unit is designed to be as simple as possible. Please view the minute long video below.
Catalog No. NC2259355 Supplier Euthanex Supplier No. EZDOCKNGM
Fits Allentown Nexgen Max cages only. The EZ-Dock combines the best features of all our systems. A built-in flow circuit and times are already preset for the specific cage type to ensure AMVA compliancy. Plug in, connect the provided gas line and the system is ready for use! Once the cage is placed in dock properly. The cycles begin and shut offs when time has elapsed. The unit is designed to be as simple as possible. Please view the minute long video below.