Learn More
Apexbio Technology LLC EZ Cap EGFP mRNA (m1Ψ) 5x1mg (1mg/mL)
SDP
Catalog No. 503107094
Encompass_Preferred
Enhanced green fluorescent protein (EGFP) is a commonly used reporter molecule that emits intense and bright green fluorescence upon excitation EZ CapTM EGFP mRNA (m1 ) is provided at a concentration of approximately 1 mg/mL It features a Cap 1 structure generated through co-transcriptional capping which is more suitable for mammalian systems and offers higher transcription efficiency compared to Cap 0 structures (ARCA and mCap) Incorporation of N1-Methylpseudo-UTP (m1 ) and a poly (A) tail helps suppress RNA-mediated innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo Additionally the poly (A) tail plays a crucial role in enhancing the efficiency of translation initiation
Catalog No. 50-310-7094 Supplier Apexbio Technology LLC Supplier No. R10035
Enhanced green fluorescent protein (EGFP) is a commonly used reporter molecule that emits intense and bright green fluorescence upon excitation EZ CapTM EGFP mRNA (m1 ) is provided at a concentration of approximately 1 mg/mL It features a Cap 1 structure generated through co-transcriptional capping which is more suitable for mammalian systems and offers higher transcription efficiency compared to Cap 0 structures (ARCA and mCap) Incorporation of N1-Methylpseudo-UTP (m1 ) and a poly (A) tail helps suppress RNA-mediated innate immune activation and enhances mRNA stability and lifespan both in vitro and in vivo Additionally the poly (A) tail plays a crucial role in enhancing the efficiency of translation initiation