Learn More
Cayman Chemical (E/Z)-Endoxifen, Purity: ≥98% (mixture of isomers), CAS: 110025-28-0, 10 mg
SDP
Catalog No. 502986165
Encompass_Preferred
A mixture of (E)-endoxifen and (Z)-endoxifen; (Z)-endoxifen inhibits 17β-estradiol-induced increases in mRNA encoding the progesterone receptor in MCF-7 cancer cells at 100 nM; (E)-endoxifen is a potential impurity in commercial preparations of (Z)-endoxifen
Catalog No. 50-298-6165 Supplier Cayman Chemical Supplier No. 2150210 MG
A mixture of (E)-endoxifen and (Z)-endoxifen; (Z)-endoxifen inhibits 17β-estradiol-induced increases in mRNA encoding the progesterone receptor in MCF-7 cancer cells at 100 nM; (E)-endoxifen is a potential impurity in commercial preparations of (Z)-endoxifen