Learn More
MSC American Specialties Top Fill Multi-Feed Kit, 6 L, 1 ea
Catalog No. 502818514
Encompass
The EZ FILL™ Top Fill Multi-Feed Kit holds 203 oz (6 L) of liquid soap, supplying up to six vanity-mounted automatic soap dispensers. It features a translucent, leak-proof plastic tank for easy soap level monitoring, situated under the counter. The tank connects to soap distribution tubing and a top fill port via a flexible hose. The top fill port and cover are chrome-plated brass.
  • Utilizes infrared sensors to feed up to six automatic soap dispensers, each controlled by its own peristaltic pump.
  • Top-fill system allows refilling up to six soap dispensers with a single pour into one reservoir, simplifying maintenance.
  • Single remote controls all dispensers, offering flexibility with four presets.
  • Holds 6 L of liquid soap to supply up to six individual vanity-mounted automatic soap dispenser units.
Catalog No. 50-281-8514 Supplier MSC Supplier No. 11775244
The EZ FILL™ Top Fill Multi-Feed Kit holds 203 oz (6 L) of liquid soap, supplying up to six vanity-mounted automatic soap dispensers. It features a translucent, leak-proof plastic tank for easy soap level monitoring, situated under the counter. The tank connects to soap distribution tubing and a top fill port via a flexible hose. The top fill port and cover are chrome-plated brass.
  • Utilizes infrared sensors to feed up to six automatic soap dispensers, each controlled by its own peristaltic pump.
  • Top-fill system allows refilling up to six soap dispensers with a single pour into one reservoir, simplifying maintenance.
  • Single remote controls all dispensers, offering flexibility with four presets.
  • Holds 6 L of liquid soap to supply up to six individual vanity-mounted automatic soap dispenser units.