Learn More
MSC ASI Mounting Bracket, Under Counter, 5 L, 1 ea
Catalog No. 502817421
Encompass
This mounting bracket is designed to securely hold one (1) EZ Fill™ Multi-Feed tank for automatic soap dispensing systems. Fabricated from durable 14 gauge type 304 stainless steel with a satin finish, it is suitable for under-counter or under-sink installation, supporting the full weight of liquid soap contents.
  • Wall mounted bracket for multi-feed containers
  • Designed for under-counter or under-sink installation
  • Fabricated from 14 gauge type 304 stainless steel
  • Supports 5 L multi-feed tanks
  • Compatible with EZ Fill™ vanity mounted automatic soap dispensing systems
  • Facilitates easy top-fill of multiple dispensers from one reservoir
Catalog No. 50-281-7421 Supplier MSC Supplier No. 11775350
This mounting bracket is designed to securely hold one (1) EZ Fill™ Multi-Feed tank for automatic soap dispensing systems. Fabricated from durable 14 gauge type 304 stainless steel with a satin finish, it is suitable for under-counter or under-sink installation, supporting the full weight of liquid soap contents.
  • Wall mounted bracket for multi-feed containers
  • Designed for under-counter or under-sink installation
  • Fabricated from 14 gauge type 304 stainless steel
  • Supports 5 L multi-feed tanks
  • Compatible with EZ Fill™ vanity mounted automatic soap dispensing systems
  • Facilitates easy top-fill of multiple dispensers from one reservoir