Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma EZ-Fit™ Filtration Unit

Catalog No. EFHAW10IS Shop All MilliporeSigma Products
Change view
Click to view available options
Grid Color:
Black
White
Packaging:
Bulk
Single
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grid Color Packaging
EFHAW10IS White Single
EFHAB10BS Black Bulk
EFHAW10BS White Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EFHAW10IS Supplier MilliporeSigma Supplier No. EFHAW10IS
Only null left
Add to Cart
Add to Cart

0.45 μm MCE membrane with 100 mL funnel, suitable for solid cultivation

The EZ-FIT™ filtration unit is a single-use filtration device made of gridded, mixed cellulose esters membrane with a pink base. It provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis.

The filtration units are e-beam irradiated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
For Use With (Equipment) EZ-Fit Filtration Manifold
Material Styrene-Butadiene
Material (Membrane) MCE
Sample Volume 100 mL
Sterility Sterile
Grid Color White
Packaging Single
Type Filtration Unit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.