Learn More
GENTECH SOLUTIONS LLC EZ-GRIP Disposable Watch Glass, 100mL
Supplier:  GENTECH SOLUTIONS LLC 1100101012
Encompass_Preferred
EliteChem Our innovative EZ-Grip Watch Glass™ line features a patent-pending, larger, ergonomic tab that enhances handling compared to other disposable watch glasses, virtually eliminating handling mishaps. Made from metals-free polypropylene, these watch glasses are fracture-resistant and ensure no leaching into samples. Designed as single-use disposables, each is intended for a fresh part with every test. Crafted from a proprietary polypropylene blend, the EZ-Grip Watch Glass is a direct replacement for lab consumables compatible with 50ml and 100ml tubes with 27mm and 42mm diameter openings. The enhanced tab provides a secure, ergonomic grip, reducing mishandling and contamination risks. Its ribbed design also supports partial refluxing during digestion.
Catalog No. NC3728752