Learn More
Research Products International Corp UltraFlux EZ Hinge PCR Tube Strips, White Tube with Clear Caps, 125 Strips of 8 Tubes per Package
SDP
Catalog No. 50998908
Encompass
UltraFlux PCR EZ Tube Strips feature a unique hinge design that allows fast and convenient opening and closing of tubes. White color tubes maximize reflectance and enhance the signal. Optically clear caps for real-time PCR(qPCR).
Size: 0.2 ml
Certified:
RNase Free
DNase Free
Pyrogen Free
PCR Inhibitor Free
Catalog No. 50-998-908 Supplier Research Products International Corp Supplier No. 249956
UltraFlux PCR EZ Tube Strips feature a unique hinge design that allows fast and convenient opening and closing of tubes. White color tubes maximize reflectance and enhance the signal. Optically clear caps for real-time PCR(qPCR).
Size: 0.2 ml
Certified:
RNase Free
DNase Free
Pyrogen Free
PCR Inhibitor Free