Learn More
Apexbio Technology LLC EZ Cap Human PTEN mRNA 5x1mg (1mg/mL)
SDP
Catalog No. 503107320
Encompass_Preferred
PTEN mRNA encodes a a tumor suppressor gene (Phosphatase and Tensin homolog deleted on chromosome 10) that antagonizes the activity of PI3K a key enzyme involved in anti-apoptotic and pro-tumorigenic Akt activation EZ Cap Human PTEN mRNA is provided at 1 mg/mL This mRNA is synthesized through co-transcriptional capping resulting in a Cap 1 structure The addition of poly(A) tail significantly enhances mRNA stability and lifetime both in vitro and in vivo Furthermore the poly(A) tail significantly contributes to improving the efficiency of translation initiation
Catalog No. 50-310-7320 Supplier Apexbio Technology LLC Supplier No. R10255
PTEN mRNA encodes a a tumor suppressor gene (Phosphatase and Tensin homolog deleted on chromosome 10) that antagonizes the activity of PI3K a key enzyme involved in anti-apoptotic and pro-tumorigenic Akt activation EZ Cap Human PTEN mRNA is provided at 1 mg/mL This mRNA is synthesized through co-transcriptional capping resulting in a Cap 1 structure The addition of poly(A) tail significantly enhances mRNA stability and lifetime both in vitro and in vivo Furthermore the poly(A) tail significantly contributes to improving the efficiency of translation initiation