Learn More
Description
Thermo Scientific™ EZ-Link Amine-PEG3-Biotin is a biotinylation reagent that contains a 3-unit polyethylene glycol (PEG) spacer arm and a terminal primary amine for conjugation and protein labeling with EDC or other crosslinkers.
Highlights:
- Use the EDC crosslinker to couple this analog to carboxyl (-COOH) groups or 5' phosphate groups
- Water-soluble, polyethylene glycol (PEG) spacer arm
- Longer chain length for reduction of steric hindrance
- PEG spacer arm transfers hydrophilicity to final conjugate
Specifications
Specifications
| Product Type | Amine-PEG3-Biotin |
| Quantity | 50 mg |
| Content And Storage | Store at 4°C. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
| Reactive Moiety | Amine |
| Solubility | Water |
| Label or Dye | Biotin |
| Spacer | Mid-length, Pegylated |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.