Learn More
Description
Thermo Scientific™ EZ-Link Biocytin is a simple variant of biotin that contains a primary amine in its valeric acid chain, which provides the essential backbone for construction of certain long-chain and trifunctional biotinylation reagents.
Highlights:
- Useful for synthesizing long-chain biotinylation reagents (can use EDC, Part No. 22980, 22981)
- An extended spacer arm intermediate for further derivatization into reactive group-specific biotinylating reagents
- Contains both carboxyl group and primary amine, which may be modified to make trifunctional reagents like Sulfo-SBED
Specifications
Specifications
| Product Type | Biocytin |
| Quantity | 100 mg |
| Content And Storage | Store at room temperature. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Impermeant |
| Reactive Moiety | Amine, Carboxylic Acid |
| Solubility | Water |
| Label or Dye | Biotin Analog |
| Spacer | Mid-length |
| Label Type | Biotin & Analogs |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.