Learn More
Description
Thermo Scientific™ EZ-Link Hydrazide-Biotin is the shortest and simplest hydrazide-activated biotinylation reagent for labeling glycoproteins and other carbohydrate-containing compounds having oxidizable sugars or aldehydes.
Highlights:
- Carbohydrate-reactive
- Shorter spacer arm than Biotin-LC-Hydrazide
- Must be dissolved in DMSO (50mM) before adding to aqueous buffer
- Use at pH 4-6 to complete coupling reaction
- Forms hydrazone bond
Specifications
Specifications
| Product Type | Hydrazide-Biotin |
| Quantity | 100 mg |
| Content And Storage | Store at 4°C. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Permeant |
| Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
| Reactive Moiety | Amine, Hydrazide |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) |
| Label or Dye | Biotin |
| Spacer | Short |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.