Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Biotin-LC-Hydrazide

Catalog No. PI21340 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21340 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21340 Supplier Thermo Scientific™ Supplier No. 21340
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylate glycoproteins (aldehydes of oxidized sugars) with this mid-length (24.7A) hydrazide-activated biotinylation reagent.

Thermo Scientific™ EZ-Link Hydrazide-LC-Biotin is a mid-length, simple, hydrazide-activated biotinylation reagent for labeling glycoproteins and other carbohydrate-containing compounds having oxidizable sugars or aldehydes.

Biotin hydrazide labeling reagents enable simple and efficient biotin labeling of antibodies and other glycoproteins.

Highlights:

  • Carbohydrate-reactive
  • Labels immunoglobulins in the Fc region
  • Long spacer arm helps prevent steric hindrances
  • Must be dissolved in DMSO (50mM) before adding to aqueous buffer
  • Use pH 4-6 to complete coupling reaction
  • Forms hydrazone bond
TRUSTED_SUSTAINABILITY

Specifications

Product Type Biotin-LC-Hydrazide
Quantity 50 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety Amine, Hydrazide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Mid-length
Label Type Biotin & Analogs
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.