Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin, No-Weigh™ Format

Catalog No. PI21901
Change view
Click to view available options
Quantity:
50 mg
10 x 2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21901 50 mg
PIA39261 10 x 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI21901 Supplier Thermo Scientific™ Supplier No. 21901BID
Add to Cart
Add to Cart

Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.

Thermo Scientific™ EZ-Link Maleimide-PEG2-Biotin is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.

Highlights:

  • Water-soluble; contains polyethylene glycol (PEG) spacer arm
  • Reacts with sulfhydryl (-SH) groups at acidic to neutral pH
  • Transfers hydrophilic PEG spacer arm to final conjugate
  • Forms a thioether bond
TRUSTED_SUSTAINABILITY

Specifications

Product Type Maleimide-PEG2-Biotin
Quantity 50 mg
Content And Storage Store desiccated at 4°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Long, Pegylated
Label Type Biotin & Analogs
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.