Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin, No-Weigh™ Format

Catalog No. PIA39261 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
10 x 2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA39261 10 x 2 mg
PI21901 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA39261 Supplier Thermo Scientific™ Supplier No. A39261
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.

Thermo Scientific™ EZ-Link Maleimide-PEG2-Biotin is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.

  • Mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.
  • Protein labeling - biotinylate antibodies or other proteins for use in protein methods
  • Thiol-reactive - reacts with sulfhydryls (-SH), such as the side-chain of cysteine (C)
  • Maleimide-activated - perform reactions at pH 6.5 to 7.5 in buffers such as PBS
  • Pegylated - spacer arm contains a hydrophilic, 2-unit, polyethylene glycol (PEG) group
  • Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
  • Irreversible - forms permanent thioether bonds; spacer arm cannot be cleaved
  • Solubility - can be dissolved directly in aqueous buffers for labeling reactions
  • Medium length - spacer arm (total length added to target) is 29.1 angstroms
TRUSTED_SUSTAINABILITY

Specifications

Product Type Maleimide-PEG2-Biotin
Quantity 10 x 2 mg
Content And Storage Store desiccated at 4°C.
Labeling Method Chemical Labeling
Shipping Condition Qualified for Ambient or Wet Ice
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Pegylated, 29.1Å
Show More Show Less
How does a product with "No-Weigh" format reduce my concern over reagent stability?

The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.